Learn More
3M™ Secure Click™ Replaceable Particulate Filter
3M™ Secure Click™ Replaceable Particulate Filter is CE approved for environments containing certain solid and liquid particles.
Brand: 3M™ 7100197285
Additional Details : Weight : 0.21000kg
Description
Use our filters with 3M™ Secure Click™ Respirators 800 Series, 3M™ Secure Click™ Cartridges D8000 Series and 3M™ Filter Retainer D701.Specifications
CE Approved | |
P2R | |
D7935 |
Secure Click™ | |
Replacement Filter |