Learn More
Tocris Bioscience™ D609
GREENER_CHOICE
Product Code. 18045952
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18045952 10 mg 10mg
18094152 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18045952 Supplier Tocris Bioscience™ Supplier No. 1437/10
  • ONLINE EXCLUSIVE PRICE
    229.00 €
    216.30 € / 10mg
    Save 12.70 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Selective PC-PLC inhibitor
Selective competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor (Ki = 6.4 μM); antiviral and antitumor agent. Also inhibits sphingomyelin synthase. Suppresses LPS- and IFNγ-induced NO production (IC50 = 20 mg/ml) and blocks oxidative glutamate toxicity in nerve cells. Antioxidant in vivo.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83373-60-8
Molecular Formula C11H18KOS2
Specifications
Quantity 10 mg
Formula Weight 266.46
Purity >98%
Molecular Weight (g/mol) 269.482
Chemical Name or Material D609