missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ D4-GDI Polyclonal Antibody
GREENER_CHOICE

Product Code. 16530302
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16530302 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16530302 Supplier Invitrogen™ Supplier No. PA597469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

D4-GDI Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D4-GDI
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ARHGDIB
Gene Accession No. P52566
Gene Alias ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2
Gene Symbols ARHGDIB
Host Species Rabbit
Immunogen Recombinant Human Rho GDP-dissociation inhibitor 2 protein (2-86AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.