missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D2HGDH Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92309-0.1ml
This item is not returnable.
View return policy
Description
D2HGDH Polyclonal antibody specifically detects D2HGDH in Mouse, Rat samples. It is validated for Western Blot
Specifications
| D2HGDH | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| D-2-hydroxyglutarate dehydrogenase, EC 1.1.99, EC 1.1.99.-, FLJ42195, MGC25181, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 322-521 of human D2HGDH (NP_689996.4). GRHLHLASPVQESPFYVLIETSGSNAGHDAEKLGHFLEHALGSGLVTDGTMATDQRKVKMLWALRERITEALSRDGYVYKYDLSLPVERLYDIVTDLRARLGPHAKHVVGYGHLGDGNLHLNVTAEAFSPSLLAALEPHVYEWTAGQQGSVSAEHGVGFRKRDVLGYSKPPGALQLMQQLKALLDPKGILNPYKTLPSQA | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 728294 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction