missing translation for 'onlineSavingsMsg'
Learn More

D-Ribose, TRC

Product Code. 30010995
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010995 250 g 250g
30138322 5 g 5g
30133486 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010995 Supplier Toronto Research Chemicals Supplier No. TRCR417500250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-69-1
Formula Weight 150.0528
InChI Formula InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5-/m0/s1
IUPAC Name (2R,3R,4R)-2,3,4,5-tetrahydroxypentanal
Molecular Formula C5 H10 O5
Molecular Weight (g/mol) 150.13
SMILES OC[C@@H](O)[C@@H](O)[C@@H](O)C=O
Synonym D-Ribose, Ribose, D- (8CI), Ribose, D-(-)-Ribose
Chemical Name or Material D-(-)-Ribose

Specifications

Quantity 250 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.