Learn More
D-Ribonolactone 2,3-Cyclohexyl Ketal, TRC
Specifications
Specifications
| Chemical Name or Material | D-Ribonolactone 2,3-Cyclohexyl Ketal |
| CAS | 27304-20-7 |
| Molecular Formula | C11H16O5 |
| Quantity | 100 mg |
| Synonym | 2,3-O-Cyclohexylidene-D-ribono-1,4-lactone, 2,3-O-Cyclohexylidene-D-ribonic Acid gamma-lactone |
| InChI Formula | InChI=1S/C11H16O5/c12-6-7-8-9(10(13)14-7)16-11(15-8)4-2-1-3-5-11/h7-9,12H,1-6H2/t7-,8-,9-/m1/s1 |
| SMILES | OC[C@H]1OC(=O)[C@@H]2OC3(CCCCC3)O[C@H]12 |
| IUPAC Name | (3aR,6R,6aR)-6-(hydroxymethyl)spiro[6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxole-2,1'-cyclohexane]-4-one |
| Molecular Weight (g/mol) | 228.24 |
| Formula Weight | 228.0998 |