missing translation for 'onlineSavingsMsg'
Learn More

D-Proline, 99%

Product Code. p-7080935
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483444 1 g 1g
11493444 5 g 5g
11403454 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483444 Supplier Thermo Scientific Alfa Aesar Supplier No. A10682.03
Add to basket
This item is not returnable. View return policy

CAS: 344-25-2 | C5H9NO2 | 115.132 g/mol

D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.

Solubility
Soluble in water (1620 g/L at 20°C), and acetone (slightly). Insoluble in ethanol.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00064317
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
PubChem CID 8988
ChEBI CHEBI:16313
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Melting Point 213°C to 215°C (decomposition)
Beilstein 80811
Solubility Information Soluble in water (1620g/L at 20°C),and acetone (slightly). Insoluble in ethanol.
Density 1.36
Quantity 1 g
Formula Weight 115.13
Optical Rotation +84° (c=1 in water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material D-Proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.