Learn More
D-Phenylalanine-d5, TRC
Product Code. 30179361
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179361 250 mg 250mg
30179441 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179361 Supplier Toronto Research Chemicals Supplier No. TRCP319412250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 362049-55-6
Molecular Formula C9 D5 H6 N O2
Molecular Weight (g/mol) 170.22
Synonym (2R)-2-Amino-3-phenylpropanoic Acid-d5, (R)-3-Phenyl-2-aminopropanoic Acid-d5, (R)-Phenylalanine-d5, D-(+)-Phenylalanine-d5, D-alpha-Amino-beta-phenylpropionic Acid-d5, Endorphenyl-d5, NSC 25005-d5, D-Phenylalanine-D5 (phenyl-D5), D-Phenylalanine-2,3,4,5,6-d5, D-Phenyl-d5-alanine (9CI), D-Phenylalanine-(phenyl-d5), D-(2,3,4,5,6-2H5)Phenylalanine, D-[2H5]Phenylalanine, d5-ring-D-Phenylalanine, (2R)-2-Amino-3-(phenyl-d5)propanoic acid, D-Phenyl-d5-alanine
IUPAC Name (2R)-2-amino-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid
SMILES [2H]c1c([2H])c([2H])c(C[C@@H](N)C(=O)O)c([2H])c1[2H]
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m1/s1/i1D,2D,3D,4D,5D
Formula Weight 170.1104
Chemical Name or Material D-Phenylalanine-d5