Learn More
Thermo Scientific Chemicals D-(-)-Norleucine, 99%
Product Code. 11358065
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358065 5 g 5g
11348065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11358065 Supplier Thermo Scientific Chemicals Supplier No. L08257.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 327-56-0 | C6H13NO2 | 131.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 327-56-0
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00008099
InChI Key LRQKBLKVPFOOQJ-RXMQYKEDSA-N
Synonym d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine
PubChem CID 456468
ChEBI CHEBI:42101
IUPAC Name (2R)-2-aminohexanoic acid
SMILES CCCC[C@@H](N)C(O)=O
Specifications
Melting Point >300°C
Beilstein 1721749
Merck Index 14,6706
Quantity 5 g
Formula Weight 131.18
Optical Rotation −23° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-(-)-Norleucine