Learn More
D-Mannose-UL-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | D-Mannose-UL-13C6 |
| CAS | 287100-74-7 |
| Molecular Formula | 13C6H12O6 |
| Quantity | 100 mg |
| Synonym | D-Mannose-Ul 13C6 |
| InChI Formula | InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1 |
| SMILES | O[13C@H]1[13C@H](O)[13C@H](O)[13CH](O)O[13C@@H]1[13CH2]O |
| Molecular Weight (g/mol) | 186.11 |
| Formula Weight | 186.0835 |