missing translation for 'onlineSavingsMsg'
Learn More

D,L-Azetidine-2-carboxylic Acid-d4, TRC

Product Code. 30334840
Change view
Click to view available options
Quantity:
1 mg
25 mg
100 mg
Unit Size:
100mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334840 25 mg 25mg
30332621 100 mg 100mg
30331905 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30334840 Supplier Toronto Research Chemicals Supplier No. TRCA81249225MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D,L-Azetidine-2-carboxylic Acid-d4
Molecular Formula C4 D4 H3 N O2
Quantity 25 mg
Synonym (+)-2-Azetidinecarboxylic Acid-d4, Azetidine Carboxylic Acid-d4
InChI Formula InChI=1S/C4H7NO2/c6-4(7)3-1-2-5-3/h3,5H,1-2H2,(H,6,7)/i1D2,2D2
SMILES [2H]C1([2H])NC(C(=O)O)C1([2H])[2H]
IUPAC Name 3,3,4,4-tetradeuterioazetidine-2-carboxylic acid
Molecular Weight (g/mol) 105.129
Formula Weight 105.073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.