Learn More
D-Homoarginine Hemisulfate, TRC
Specifications
Specifications
| Chemical Name or Material | D-Homoarginine Hemisulfate |
| CAS | 32733-73-6 |
| Molecular Formula | 2(C7H16N4O2) • (H2O4S) |
| Quantity | 250 mg |
| Synonym | N6-(Aminoiminomethyl)-D-lysine Hemisulfate Salt, (R)-2-Amino-6-guanidinohexanoic Acid Hemisulfate Salt |
| InChI Formula | InChI=1S/C7H16N4O2.H2O4S/c8-5(6(12)13)3-1-2-4-11-7(9)10;1-5(2,3)4/h5H,1-4,8H2,(H,12,13)(H4,9,10,11);(H2,1,2,3,4)/t5-;/m1./s1 |
| SMILES | N[C@H](CCCCNC(N)=N)C(O)=O.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 2188.239808 |
| Formula Weight | 474.222 |