missing translation for 'onlineSavingsMsg'
Learn More

D-Glucose, MedChemExpress

Product Code. 16422159
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16422159

Brand: Medchem Express HYB0389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. D-Glucose is a carbohydrate sweetener and critical components of the general metabolism, and serve as critical signaling molecules in relation to both cellular metabolic status and biotic and abiotic stress response.

D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. D-Glucose is a carbohydrate sweetener and critical components of the general metabolism, and serve as critical signaling molecules in relation to both cellular metabolic status and biotic and abiotic stress response.

Specifications

Chemical Name or Material D-Glucose
Physical Form Solid
CAS 50-99-7
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C6H12O6
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym Glucose D-(+)-Glucose Dextrose
Solubility Information H2O : ≥ 50 mg/mL (277.53 mM)
SMILES O=C[C@@H]([C@H]([C@@H]([C@@H](CO)O)O)O)O
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 96.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.