missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%

Product Code. 11452957
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11452957

Brand: Thermo Scientific Chemicals A15532.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Glucosamine hydrochloride, CAS # 3416-24-8, is the salt of the amino sugar D-Glucosamine, used for chitin quantitation and other biochemical study.

Novel application of glucosamine is to prepare medical agent for treating vertigo. It is used as food ingredients and additives, raw material for anti cancer and antibiotic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D-Glucosamine is an abundant monosaccharide amino sugar that acts as a building block in the synthesis of glycosylated proteins and lipids

• D-Glucosamine is the precursor at the start of the hexosamine pathway that is essential to the formation of common proteoglycans

• D-Glucosamine combines with chondroitin sulfate in cartilage and is found as a monomer in chitin

Application

• D-Glucosamine can be used as a biochemical standard or for cell stimulation along with glucose in cell culture

• It may be useful in the laboratory study of osteoarthritis and diabetes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5·HCl
MDL Number MFCD00067674

Specifications

Melting Point ∼192°C (decomposition)
Quantity 1000 g
Beilstein 4157370
Merck Index 14,4458
Solubility Information Soluble in water.
Optical Rotation +71.5° (c=1 in Water, 20h)
Formula Weight 215.64
Percent Purity ≥98%
Chemical Name or Material D-Glucosamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.