missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ [D-Arg0, Hyp3, Thi5,8, D-Phe7]-Adamantaneacetyl Bradykinin

Product Code. 15430637
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15430637

Brand: Thermo Scientific™ J66709.M^

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name [N-Admantanecarbonyl-D-Arg0-Hyp3, Thi2\5, 8, D-Phe7] Bradykinin
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1455.72
Product Type [D-Arg0, Hyp3, Thi5,8, D-Phe7]-Admantanecarbonyl Bradykinin
Quantity 1 mg
Regulatory Status RUO - research use only
Sensitivity Light sensitive
Sequence N-Adamantanecarbonyl-D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-D-Phe-Thi-Arg

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.