Learn More
D-Arabinono-1,4-lactone, TRC
Specifications
Specifications
| Chemical Name or Material | D-Arabinono-1,4-lactone |
| CAS | 2782-09-4 |
| Molecular Formula | C5H8O5 |
| Quantity | 1 g |
| Synonym | D-Arabinonic Acid Gamma-lactone |
| InChI Formula | InChI=1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3-,4+/m1/s1 |
| SMILES | OC[C@H]1OC(=O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one |
| Molecular Weight (g/mol) | 148.11 |
| Formula Weight | 148.0372 |