Learn More
D-Allose, TRC
Product Code. 30179556
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179556 1 g 1g
30179276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179556 Supplier Toronto Research Chemicals Supplier No. TRCA5480051G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2595-97-3
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym (2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanol, D-Allose
IUPAC Name (2R,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@@H](O)[C@@H](O)[C@@H](O)[C@@H](O)C=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5-,6+/m0/s1
Formula Weight 180.0634
Chemical Name or Material D-(+)-Allose