missing translation for 'onlineSavingsMsg'
Learn More

D-64131, MedChemExpress

Product Code. p-200099442
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16442657

Brand: Medchem Express HY15482

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-64131 is an orally active tubulin inhibitor, with an IC50 of 0.53 μM for tubulin polymerization. D-64131 has antimitotic activity. D-64131 can be used for cancer research.

D-64131 is an orally active tubulin inhibitor, with an IC50 of 0.53 μM for tubulin polymerization. D-64131 has antimitotic activity. D-64131 can be used for cancer research.

Specifications

Chemical Name or Material D-64131
CAS 74588-78-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13NO2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (397.96 mM)
SMILES O=C(C(N1)=CC2=C1C=CC(OC)=C2)C3=CC=CC=C3
Molecular Weight (g/mol) 251.28
Formula Weight 251.28
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.