missing translation for 'onlineSavingsMsg'
Learn More

CZC-8004, MedChemExpress

Product Code. p-200097598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16497127

Brand: Medchem Express HY111138

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CZC-8004 is a pan-kinase inhibitor and binds a range of tyrosine kinases, including ABL kinase.

CZC-8004 is a pan-kinase inhibitor and binds a range of tyrosine kinases, including ABL kinase.

Specifications

Chemical Name or Material CZC-8004
Physical Form Solid
CAS 916603-07-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H16FN5
For Use With (Application) Cancer-Kinase/protease
Synonym CZC-00008004
Solubility Information DMSO : 77.5 mg/mL (250.53 mM; Need ultrasonic)
SMILES FC1=CN=C(NC2=CC=C(CN)C=C2)N=C1NC3=CC=CC=C3
Molecular Weight (g/mol) 309.34
Formula Weight 309.34
Percent Purity 99.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.