missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
310.00 € - 507.00 €
Specifications
| Antigen | Cytomegalovirus gB |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612769
|
Novus Biologicals
NBP3-06076-100ul |
100 μg |
507.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683187
|
Novus Biologicals
NBP3-06076-50ul |
50 μg |
310.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytomegalovirus gB Polyclonal antibody specifically detects Cytomegalovirus gB in Cytomegalovirus samples. It is validated for ELISASpecifications
| Cytomegalovirus gB | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Human Cytomegalovirus gB (Accession#: AAA45920.1) extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Cytomegalovirus (CMV) | |
| CMV, CMV glycoprotein B, CMV glycoprotein GP55, Cytomegalovirus glycoprotein GP55, gB, Glycoprotein B, Glycoprotein GP55, HCMV, HCMV glycoprotein B, HCMV glycoprotein GP55, HHV 5 glycoprotein B, HHV 5 glycoprotein GP55, HHV5 glycoprotein B, HHV5 glycoprotein GP55, Human herpesvirus 5 glycoprotein B, Human herpesvirus 5 glycoprotein GP55, UL55 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title