missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cytomat™ 2 C425-LiN Automated Incubator, Copper

Product Code. p-9053103 Shop All Thermo Scientific Products
Click to view available options
Voltage:
230 V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18887681

Brand: Thermo Scientific™ 51033034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Temperature Range: Ambient +4°C to 25°C; Relative Humidity: <90%

Ensure a safe, reproducible process, unsurpassed temperature uniformity and stability with the next generation of mid-range Thermo ScientificTM CytomatTM 2 C4xx-LiN series automated incubators for incubation/storage of cell culture applications.

These Cytomat incubators are available in various temperature ranges and gate positions for the hand-off to the lab automation robot. The device can be seamlessly integrated with a mean access time of 12 sec.

With a fully automated thermal decontamination and optional controlled humidity, the Cytomat 2 C4xx-LiN series provides a new standard in significantly reducing the risk of contamination.

The Cytomat 2 C4xx-LiN series of automated incubators provide an ideal safe environment for storage and incubation of assays. This series of mid-range incubators offer a choice of temperature ranges (4 to 70°C), high humidity up to 95% r.H. stable CO2 conditions, and automated ContraCon decontamination routine without harmful gases.

The external cooling and heating unit is designed to allow the user to create a customized working space for easy system integration into any automated assay system.

Available options

  • Integrated barcode reader (1D/2D) enables sample tracking, barcode transmission, and inventory management.
  • P3-Plate detection in stacker helps to eliminate labware handling errors.
  • HydraSmart Technology for precise humidity control without heat extraction and external water tank to avoid contamination.
  • Right speed setting for the right application with individual speed setting per plate and adjustable incremental speed settings (Slow/Medium/Fast).
  • Flexible positioning inside your lab with 6 possible gate positions including under bench in a single body.
  • Solid Copper inner chamber for natural contamination prevention.
  • Tower Shaker (ToS) for TRUE orbital shaking of applications requiring sample agitation and cells that need to be kept in suspension.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Access Time 12 sec.
Chamber Material Copper
Holds 56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well
Relative Humidity <90%
Temperature Range (Metric) Ambient +4° to 25°C
Type Automated Incubator
Features Incubation and storage unit with temperature range above; Humidity up to 90% r.H.; Includes decontamination routine-ContraCon
For Use With (Application) Incubation/storage for cell culture applications 4 to 25°C
Model Cytomat 2 C425-LiN 230V 50Hz
Width (English) 22.5 in.
Width (Metric) 572mm
Height (English) 35.6 in.
Height (Metric) 905mm
Depth (English) 20.1 in.
Depth (Metric) 511mm
Weight (Metric) 90kg
Electrical Requirements 230V 50 Hz
Voltage 230 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.