missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cytokeratin, LMW Monoclonal specifically detects Cytokeratin, LMW in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit, Reptile samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | Cytokeratin, LMW |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AE-1 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Alias | Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39 |
| Gene Symbols | KRT77 |
| Host Species | Mouse |
| Immunogen | Human epidermal keratin (Uniprot: Q7Z794) |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?