missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10405-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| Cytohesin 2 | |
| Polyclonal | |
| Unconjugated | |
| CYTH2 | |
| ARF exchange factor, ARNO, CTS18, CTS18.1, CYTH2, cytohesin 2, Protein ARNO, PSCD2, PSCD2L, SEC7L, Sec7p L, Sec7p like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116692, 19158, 9266 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P63034, P63035, Q99418 | |
| Cyth2 | |
| Cytohesin 2 Fusion Protein Ag0687 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction