missing translation for 'onlineSavingsMsg'
Learn More

Cytarabine, MedChemExpress

Product Code. 16491359
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 16491359

Brand: Medchem Express HY13605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cytarabine, a nucleoside analog, causes S phase cell cycle arrest and inhibits DNA polymerase. Cytarabine inhibits DNA synthesis with an IC50 of 16 nM. Cytarabine has antiviral effects against HSV. Cytarabine shows anti-orthopoxvirus activity.

Cytarabine, a nucleoside analog, causes S phase cell cycle arrest and inhibits DNA polymerase. Cytarabine inhibits DNA synthesis with an IC50 of 16 nM. Cytarabine has antiviral effects against HSV. Cytarabine shows anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Cytarabine
Physical Form Powder
CAS 147-94-4
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H13N3O5
For Use With (Application) Cancer-Kinase/protease
Synonym Cytosine β-D-arabinofuranoside Cytosine Arabinoside Ara-C
Solubility Information H2O : 48 mg/mL (197.35 mM; Need ultrasonic) ∣DMSO : 17.3 mg/mL (71.13 mM; Need ultrasonic and warming)
SMILES O=C1N=C(N)C=CN1[C@H]2[C@H]([C@@H]([C@@H](CO)O2)O)O
Molecular Weight (g/mol) 243.22
Formula Weight 243.22
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.