missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYPC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10287-2-AP-20UL
This item is not returnable.
View return policy
Description
PPIC is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A.Specifications
| CYPC | |
| Polyclonal | |
| Unconjugated | |
| PPIC | |
| Cyclophilin C, CYPC, PPIase C, PPIC, Rotamase C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19038, 291463, 5480 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P30412, P45877 | |
| Ppic | |
| CYPC Fusion Protein Ag0209 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction