missing translation for 'onlineSavingsMsg'
Learn More

CYP2A6 Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene

Product Code. 17510122
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17510122 30 μL 30µL
17590112 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17510122 Supplier Origene Technologies Supplier No. TA503827S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CYP2A6 Monoclonal antibody specifically detects CYP2A6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CYP2A6
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A8
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ADIPOQ
Gene Accession No. Q15848
Gene Alias 1,4-cineole 2-exo-monooxygenase; Coumarin 7-hydroxylase; CPA6; CYP; CYP2A; CYP2A3; CYP2A6; CYPIIA6; cytochrome P450 2A6; cytochrome P450 family 2 subfamily A member 6; cytochrome P450 IIA3; Cytochrome P450(I); cytochrome P450, family 2, subfamily A, polypeptide 6; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6; flavoprotein-linked monooxygenase; P450; P450 IIA3; P450(I); P450C2A; P450PB; xenobiotic monooxygenase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.