missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP27B1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92637-0.02ml
This item is not returnable.
View return policy
Description
CYP27B1 Polyclonal antibody specifically detects CYP27B1 in Human, Rat samples. It is validated for Western Blot
Specifications
| CYP27B1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| 1alpha(OH)ase, 25 hydroxyvitamin D3-1-alpha hydroxylase, 25-hydroxyvitamin D(3) 1-alpha-hydroxylase, 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial, 25-OHD-1 alpha-hydroxylase, Calcidiol 1-monooxygenase, CP2B, CYP1, CYP1ALPHA, CYP27B, Cytochrome p450 27B1, Cytochrome P450 subfamily XXVIIB polypeptide 1, cytochrome P450, family 27, subfamily B, polypeptide 1, cytochrome P450, subfamily XXVIIB (25-hydroxyvitamin D-1-alpha-hydroxylase), Cytochrome P450C1 alpha, Cytochrome P450VD1-alpha, EC 1.14.13.13, P450c1, PDDR, polypeptide 1, VD3 1A hydroxylase, VDD1, VDDR, VDDRI, VDR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-195 of human CYP27B1 (NP_000776.1). LGYREYHSARRSLADIPGPSTPSFLAELFCKGGLSRLHELQVQGAAHFGPVWLASFGTVRTVYVAAPALVEELLRQEGPRPERCSFSPWTEHRRCRQRACGLLTAEGEEWQRLRSLLAPLLLRPQAAARYAGTLNNVVCDLVRRLRRQRGRGTGPPALVRDVAGEFYKFGL | |
| 0.02 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1594 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction