missing translation for 'onlineSavingsMsg'
Learn More

CYP27A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00 € - 589.00 €

Specifications

Antigen CYP27A1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18469961
View Documents
Novus Biologicals
NBP2-32433-25ul
25 μL
280.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18136852
View Documents
Novus Biologicals
NBP2-32433
0.1 mL
589.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CYP27A1 Polyclonal specifically detects CYP27A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CYP27A1
Polyclonal
Rabbit
Stem Cell Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
1593
This antibody was developed against a recombinant protein corresponding to amino acids: RQRSLEEIPRLGQLRFFFQLFVQGYALQLHQLQVLYKAKYGPMWMSYLGPQMHVNLASAPLLEQVMRQEGKYPVRNDMELWKEHRDQHDLTYGPFTTEGHHWYQLRQALNQRLLKPAEAALYTDAFNEVIDDFMTRLD
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
12-alpha-triol 27-hydroxylase, 7-alpha, cholestanetriol 26-monooxygenase, CP27,5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase, CTX5-beta-cholestane-3-alpha, CYP275-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase, Cytochrome P450 27, cytochrome P450, family 27, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinousxanthomatosis), polypeptide 1, Cytochrome P-450C27/25, EC 1.14.13.15, sterol 26-hydroxylase, mitochondrial, Sterol 27-hydroxylase, Vitamin D(3) 25-hydroxylase
CYP27A1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.