missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cynomolgus Monkey VSIG4 (XP_005593850) VersaClone cDNA

Product Code. 18752308
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752308 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18752308 Supplier R&D Systems™ Supplier No. RDC2579
  • ONLINE EXCLUSIVE PRICE
    347.14 €
    309.75 € / 10µg
    Save 37.39 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • VSIG4 (V-set and immunoglobulin domain containing 4), also known as CRIg and Z39IG, is a 45 kDa, type I transmembrane protein of the B7 family within the Ig superfamily that is expressed only in tissue-resident macrophages.
  • The gene is located on the X chromosome.
  • The human VSIG4 cDNA encodes 399 amino acids (aa) including a 19 aa signal sequence, a 264 aa extracellular domain (ECD) containing a V-type and a C2-type Ig domain, a 21 aa transmembrane domain and a 95 aa cytoplasmic domain.
  • The human VSIG4 ECD shares 84% aa identity with canine VSIG4.
  • Within the IgV domain, it shares 90%, 80% and 78% aa identity with bovine, mouse and rat VSIG4, respectively; these animals lack the C2-type domain.
  • Splice isoforms of 321, 305, 272, 201 and 199 aa lack all or part of the cytoplasmic domain, the C2-type Ig domain and/or the transmembrane domain.
  • VSIG4 is specifically expressed on macrophages in the thymic medulla, peritoneum, alveoli, synovia, adipose and heart, liver Kupffer cells, placental Hofbauer cells, and atherosclerotic foam cells. It is absent on infiltrating macrophages.
  • VSIG4 is a complement receptor that binds C3b and iC3b fragments, internalizes them to recycling endosomes, and is recycled to the cell surface.
  • It contributes significantly to innate immunity by binding and phagocytosis of complement-opsonized invading pathogens.
  • Binding of either native or recombinant soluble VSIG4 to C3b inhibits complement amplification through the alternative, but not classical, pathway.
  • VSIG4 is also a negative regulator of mouse and human T cell activation.
  • Although VSIG4 engagement may activate NF-kappa-B and thus be pro-inflammatory in some cases, many of its activities are important in resolving, rather than initiating, inflammation.
  • The VSIG4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene VSIG4
Accession Number XP_005593850
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1228 bp
Gene ID (Entrez) 11326
Gene Alias CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Product Line R&D Systems
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.