missing translation for 'onlineSavingsMsg'
Learn More

CYM-5541, MedChemExpress

Product Code. p-200097318
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496077

Brand: Medchem Express HY101419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CYM-5541 (ML249) is an selective and allosteric S1P3 receptor agonist with an EC50 between 72 and 132 nM.

CYM-5541 (ML249) is an selective and allosteric S1P3 receptor agonist with an EC50 between 72 and 132 nM.

Specifications

Chemical Name or Material CYM-5541
Physical Form Solid
CAS 945128-26-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H28N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym ML249
Solubility Information DMSO : 25 mg/mL (79.00 mM; Need ultrasonic)
SMILES O=C(C1=NOC(C2CC2)=C1)N(C3CCCCC3)C4CCCCC4
Molecular Weight (g/mol) 316.44
Formula Weight 316.44
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.