missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclophilin F Recombinant Rabbit Monoclonal Antibody (23GB2140), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553232
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death.
Specifications
| Cyclophilin F | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29117, P30405, Q99KR7 | |
| Ppif | |
| A synthesized peptide derived from human Cyclophilin F (1-66AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2140 | |
| Unconjugated | |
| Ppif | |
| AW457192; cyclophilin 3; cyclophilin D; Cyclophilin F; CYP3; CypD; CyP-D; CyP-F; CyP-M; mitochondrial cyclophilin; mitochondrial Cyclophilin D; mitochondrial peptidyl-prolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase F; peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; peptidylprolyl isomerase F; peptidylprolyl isomerase F (cyclophilin F); PPIase; PPIase F; PPIF; Rotamase F | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10105, 105675, 282819 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction