missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylbenzene, 97+%

Product Code. p-7023181
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11417797

Brand: Thermo Scientific Alfa Aesar A16320.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 827-52-1 | C12H16 | 160.26 g/mol

Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-52-1
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00001451
InChI Key IGARGHRYKHJQSM-UHFFFAOYSA-N
Synonym phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
PubChem CID 13229
IUPAC Name cyclohexylbenzene
SMILES C1CCC(CC1)C1=CC=CC=C1

Specifications

Melting Point 5°C to 6°C
Density 0.945
Boiling Point 239°C to 240°C
Flash Point 98°C (208°F)
Odor Mild
Refractive Index 1.526
Quantity 100 g
UN Number UN3082
Beilstein 1906803
Solubility Information Immiscible with water.
Formula Weight 160.26
Percent Purity ≥97%
Chemical Name or Material Cyclohexylbenzene
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.