missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl sulfide, 97%

Product Code. p-7043964
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11362276

Brand: Thermo Scientific Alfa Aesar L03712.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7133-37-1 | C7H14S | 130.25 g/mol

Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in water 173.5 mg/L @ 25°C, alcohol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7133-37-1
Molecular Formula C7H14S
Molecular Weight (g/mol) 130.25
MDL Number MFCD00039460
InChI Key QQBIOCGHCKNYGP-UHFFFAOYSA-N
Synonym cyclohexyl methyl sulfide, methylthio cyclohexane, sulfide, cyclohexyl methyl, cyclohexane, methylthio, methylthiocyclohexane, methylcyclohexylsulfide, cyclohexylmethylsulphide, cyclohexyl methylsulfide, methyl cyclohexyl sulfide, methylsulfanyl cyclohexane
PubChem CID 81553
SMILES CSC1CCCCC1

Specifications

Melting Point -80°C
Density 0.944
Boiling Point 89°C to 90°C (35 mmHg)
Flash Point 61°C (142°F)
Odor Unpleasant
Refractive Index 1.494
Quantity 5 g
UN Number UN3334
Beilstein 1919309
Solubility Information Soluble in water 173.5mg/L @ 25°C,alcohol.
IUPAC Name (methylsulfanyl)cyclohexane
Formula Weight 130.25
Percent Purity 97%
Chemical Name or Material Cyclohexyl methyl sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.