Learn More
Cyclo(Leu-Gly), TRC
Product Code. 30363453
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363453 100 mg -
30357244 50 mg 50mg
30362201 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30363453 Supplier Toronto Research Chemicals Supplier No. TRCC984485100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5845-67-0
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
Synonym Cyclo(Gly-L-Leu), (S)-3-Isobutylpiperazine-2,5-dione, (3S)-3-(2-Methylpropyl)piperazine-2,5-dione, L-3-Isobutyl-2,5-piperazinedione, Cyclo(glycyl-L-leucyl), Cyclo(L-leucylglycine), Cyclo(L-leucylglycyl), L-Leucylglycine Diketopiperazine
IUPAC Name (3S)-3-(2-methylpropyl)piperazine-2,5-dione
SMILES CC(C)C[C@@H]1NC(=O)CNC1=O
Specifications
InChI Formula InChI=1S/C8H14N2O2/c1-5(2)3-6-8(12)9-4-7(11)10-6/h5-6H,3-4H2,1-2H3,(H,9,12)(H,10,11)/t6-/m0/s1
Quantity 100 mg
Formula Weight 170.1055
Shipping Condition Room Temperature
Chemical Name or Material Cyclo(Leu-Gly)