missing translation for 'onlineSavingsMsg'
Learn More

CYB5R4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16830444 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16830444 20 μL 20µL
16820444 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16830444 Supplier Proteintech Supplier No. 126261AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).

Specifications

Antigen CYB5R4
Applications Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CYB5R4
Gene Accession No. Q3TDX8, Q68EJ0, Q7L1T6
Gene Alias b5+b5R, cb5/cb5R, CYB5R4, cytochrome b5 reductase 4, Flavohemoprotein b5/b5R, NCB5OR, RP4 676J13.1
Gene Symbols CYB5R4
Host Species Rabbit
Immunogen CYB5R4 Fusion Protein Ag3318
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171015, 266690, 51167
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.