Learn More
Cyanidin 3-O-Beta-D-Galactopyranoside Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Cyanidin-3-O-galactoside chloride (Ideain chloride) |
| CAS | 27661-36-5 |
| Molecular Formula | C21 H21 O11 . Cl |
| Quantity | 10 mg |
| Synonym | 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3-(β-D-galactopyranosyloxy)-5,7-dihydroxy-, chloride (1:1), 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3-(β-D-galactopyranosyloxy)-5,7-dihydroxy-, chloride (9CI), Idein (6CI,7CI,8CI), 3-(Galactosyloxy)-3',4',5,7-tetrahydroxyflavylium chloride, 3-O-(β-D-Galactopyranosyl)cyanidin, Cyanidin 3-O-galactoside, Cyanidin 3-O-β-D-galactopyranoside, Cyanidin 3-O-β-galactopyranoside, Cyanidin 3-galactoside, Cyanidin 3-monogalactoside, Cyanidin galactoside, Cyanindin-3-Gal, Idaein, Ideain, Ideanin, Idein chloride |
| InChI Formula | InChI=1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17+,18+,19-,21-;/m1./s1 |
| SMILES | [Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O)cc(O)cc3[o+]c2c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O |
| IUPAC Name | (2S,3R,4S,5R,6R)-2-[2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromenylium-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;chloride |
| Molecular Weight (g/mol) | 484.84 |
| Formula Weight | 484.0772 |