Learn More
MedChemExpress Cy2-SE (iodine)
Click to view available options
:
5 mg
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 643.47 g/mol |
| Solubility | DMSO : 250 mg/mL (388.52 mM; Need ultrasonic) |
| Content And Storage | -20°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 186205-33-4 |
| Synonym | Cyanine2 Succinimidyl Ester (iodine) |