missing translation for 'onlineSavingsMsg'
Learn More

CX516, MedChemExpress

Product Code. 16404298
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16404298

Brand: Medchem Express HY10933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CX516 (BDP 12) is an ampakine and acts as an AMPA receptor positive allosteric modulator for the research of Alzheimer's disease, schizophrenia and mild cognitive impairment (MCI).

CX516 (BDP 12) is an ampakine and acts as an AMPA receptor positive allosteric modulator for the research of Alzheimer's disease, schizophrenia and mild cognitive impairment (MCI).

Specifications

Chemical Name or Material CX516
Physical Form Solid
CAS 154235-83-3
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H15N3O
For Use With (Application) Neuroscience-Neurodegeneration
Synonym BDP 12
Solubility Information H2O : 100 mg/mL (414.44 mM; Need ultrasonic) ∣DMSO : ≥ 41 mg/mL (169.92 mM)
SMILES O=C(N1CCCCC1)C2=CC3=NC=CN=C3C=C2
Molecular Weight (g/mol) 241.29
Formula Weight 241.29
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.