Learn More
Invitrogen™ CX3CL1 (Fractalkine) Polyclonal Antibody, eBioscience™

Description
Description: The rabbit polyclonal antibody reacts with recombinant and natural mouse, rat, and human fractalkine (CX3CL1); the antibody was generated using E. coli-expressed chemotactic domain of human CX3CL1 as an immunogen. CX3CL1, also known as neurotactin or fractalkine, is a membrane-bound CX3C chemokine. The mature protein is part of a 397-amino acid precursor and consists of a chemokine domain of 76 amino acids, a mucin stalk of 241 amino acids, a putative transmembrane domain of 18 amino acids, and an intracellular tail of 37 amino acids. Within the chemokine domain, the first two cysteine residues are separated by 3 amino acids (CX3C). CX3CL1 message is found in high abundance in the brain, kidney, lung and heart tissues. CX3CL1 is chemotactic for monocytes and other leukocytes including NK cells and may play a role in brain inflammation. Applications Reported: This polyclonal has been reported for use in immunoblotting (WB), immunoprecipitation, and immunohistochemistry. Cross-reactivity to CX3CL1 of other species has not been determined. Applications Tested: To achieve best results, titrate antibody by dilution for each application as follows: Western Blot 1:1000-1:5000 Immunoprecipitation 1:300-1:800 Immunohistochemistry 1:100-1:500. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CX3CL1 (Fractalkine) |
| Applications | Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Cx3cl1 |
| Gene Accession No. | O35188, O55145, P78423 |
| Gene Alias | A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1 |
| Gene Symbols | Cx3cl1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.