Learn More
CWHM-12, MedChemExpress
Product Code. 16463577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16463577
Brand: Medchem Express HY18644
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CWHM-12 is a potent inhibitor of αV integrins with IC50s of 0.2, 0.8, 1.5, and 1.8 nM for αvβ8, αvβ3, αvβ6, and αvβ1.
CWHM-12 is a potent inhibitor of αV integrins with IC50s of 0.2, 0.8, 1.5, and 1.8 nM for αvβ8, αvβ3, αvβ6, and αvβ1.
Specifications
Chemical Name or Material CWHM-12
CAS 1564286-55-0
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C26H32BrN5O6
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (169.36 mM; Need ultrasonic)
SMILES O=C(CNC(C1=CC(NC2=NCC(O)CN2)=CC(O)=C1)=O)N[C@H](C3=CC(C(C)(C)C)=CC(Br)=C3)CC(O)=O
Molecular Weight (g/mol) 590.47
Formula Weight 590.47
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death