Learn More
CW-069, MedChemExpress
Product Code. 16429417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429417 5 mg 5mg
16436878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16429417 Supplier Medchem Express Supplier No. HY15857
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
MedChemExpress CW-069 is an allosteric inhibitor of microtubule motor protein HSET with an IC50 of 75 μM.
CW-069 is an allosteric inhibitor of microtubule motor protein HSET with an IC50 of 75 μM.
Specifications
Chemical Name or Material CW-069
CAS 1594094-64-0
Color White
Purity Grade Notes Research
Molecular Formula C23H21IN2O3
Quantity 5 mg
Solubility Information DMSO : ≥ 52 mg/mL (103.93 mM)
SMILES O=C(NC1=CC=C(I)C=C1C(O)=O)[C@@H](NCC2=CC=CC=C2)CC3=CC=CC=C3
Molecular Weight (g/mol) 500.33
Formula Weight 500.33
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death