Learn More
CVT-12012, MedChemExpress
Product Code. 16467097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16467097
Brand: Medchem Express HY11034
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.
CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.
Specifications
Chemical Name or Material CVT-12012
Physical Form Powder
CAS 1018675-35-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H21F3N4O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 125 mg/mL (287.75 mM; Need ultrasonic)
SMILES O=C(NCCN1C(C(C)=NC2=C1C=C(NCC3=CC=CC(C(F)(F)F)=C3)C=C2)=O)CO
Molecular Weight (g/mol) 434.41
Formula Weight 434.41
Percent Purity 99.16%
Grade Research