missing translation for 'onlineSavingsMsg'
Learn More

CVT-12012, MedChemExpress

Product Code. 16453056
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453056

Brand: Medchem Express HY11034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.

CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively.

Specifications

Chemical Name or Material CVT-12012
Physical Form Powder
CAS 1018675-35-8
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H21F3N4O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 125 mg/mL (287.75 mM; Need ultrasonic)
SMILES O=C(NCCN1C(C(C)=NC2=C1C=C(NCC3=CC=CC(C(F)(F)F)=C3)C=C2)=O)CO
Molecular Weight (g/mol) 434.41
Formula Weight 434.41
Percent Purity 99.16%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.