Learn More
Tocris Bioscience™ CVT 10216
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CVT 10216 is a potent and selective, reversible inhibitor of aldehyde dehydrogenase 2 (ALDH2) (IC50 values are 29 and 1300 nM for ALDH2 and ALDH1, respectively). Exhibits anxiolytic effects in rat models.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | CVT 10216 |
| Purity | >98% |
| Molecular Weight (g/mol) | 465.48 |
| Inhibitors | CVT 10216 |