missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CUX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116300
This item is not returnable.
View return policy
Description
CUX1 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| CUX1 | |
| Polyclonal | |
| Unconjugated | |
| CUX1 | |
| CASP; CCAAT displacement protein; CDP; CDP/Cut; CDP1; CDP2; Cdpl1; Clox; COY1; cut homolog; cut like homeobox 1; CUTL1; cut-like 1; cut-like homeobox 1; Cux; Cux/CDP; Cux/CDP homeoprotein; CUX1; Cux-1; golgi integral membrane protein 6; GOLIM6; Homeobox protein cut-like 1; homeobox protein cux-1; Kiaa4047; Nbla10317; p100; p110; p200; p75; Protein CASP; putative protein product of Nbla10317 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1523 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P39880, Q13948 | |
| CUX1 | |
| KLH conjugated synthetic peptide between 640-666 amino acids from the C-terminal region of human CUX1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction