missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ CuVer™ 2 Copper Reagent, AccuVac™ Ampules
Click to view available options
Quantity:
25 Pack
Unit Size:
Each
Description
- Fast: Just snap, fill and read - saving you valuable time
- Easy: No need to measure sample or reagents
- Accurate: Optical quality evacuated vials assure precise measurements and reagent stability
- Use them anywhere: Ampules are both reagent containers and photometric measurement cells, so no need to transport reagents or samples
Specifications
Specifications
| Analytes | Copper (Cu) |
| Format | Accuvac™ Pre-filled Ampules |
| For Use With (Application) | Copper determination |
| For Use With (Equipment) | Photometer |
| No. of Tests | 25 |
| Quantity | 25 Pack |
| Range | 0.04 to 5.00mg/L Cu |
| Test Method | Bicinchoninate |
| Type | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction