Learn More
Fisherbrand™ Cut Out Rings
With cut out section to allow easy removal of equipment.
Brand: Fisherbrand™
Additional Details : Weight : 0.22000kg
Description
- Stem length to the center of bosshead is approximately 60mm
- Constructed in mild steel with welded joints
- Zinc-plated
Specifications
Mild Steel | |
Stativring | |
Zinc Plated |
100 mm | |
Cut out section |