Learn More
Custom Primer
Product Code. 10797215
For Research Use Only. All usage must comply with product instructions.
Product Code. 10797215 Supplier Invitrogen Supplier No. 56831
  • ONLINE EXCLUSIVE PRICE
    1080.00 €
    669.60 € / Pack of 1
    Save 410.40 €
    38% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Research Use Only. All usage must comply with product instructions.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).