Learn More
Custom Primer
Product Code. 10134973
For Research Use Only. All usage must comply with product instructions.
Product Code. 10134973 Supplier Invitrogen Supplier No. 11751013
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Research Use Only. All usage must comply with product instructions.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).