Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Di-repeats
Product Code. 15507240
Change view
Click to view available options
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
10000pmol
300000pmol
80000pmol
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15507240 10,000 pmol 10000pmol
15537240 80,000 pmol 80000pmol
15537280 300,000 pmol 300000pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 15507240 Supplier Applied Biosystems™ Supplier No. 4304976
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Custom 5' Fluorescent Labeled/Unlabeled Pairs
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
TRUSTED_SUSTAINABILITY
Specifications
Description Custom 5' Labeled/Unlabeled Di-Repeats, 10000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, Choice of Delivered Scales, These Products can be used in a Variety of Applications
Product Type Custom 5' Labeled or Unlabeled Di-Repeats
Content And Storage -20°C
Quantity 10,000 pmol
For Use With (Application) Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Form Delivered either in Solution or Dried down